Assay Details
Gene Symbol: C2H3orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095395.4 | XP_001095395.2 | 731 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001095395.4 transcripts.
Set Membership: |
![]() ![]() |
