Assay Details
Gene Symbol: C2H3orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095395.4 | XP_001095395.2 | 912 | 83 | ||
XM_015130921.2 | XP_014986407.1 | 806 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
