Assay Details
| Gene Symbol: TOMM20L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143790.2 | XP_014999276.2 | 167 | 148 | ||
| XM_028851572.1 | XP_028707405.1 | 167 | 148 | |||
More Information
Set Membership: |
|

TOMM20L
Genomic Map