Assay Details
| Gene Symbol: SFTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143588.2 | XP_014999074.1 | 2688 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFTA3| Gene Symbol: SFTA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143588.2 | XP_014999074.1 | 2688 | 73 | ||