Assay Details
| Gene Symbol: TCP10L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001095991.4 | XP_001095991.2 | 108 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001095991.4 transcripts.
Set Membership: |
|

TCP10L
Genomic Map