Assay Details
| Gene Symbol: NXPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115794.2 | XP_014971280.2 | 154 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NXPE2| Gene Symbol: NXPE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115794.2 | XP_014971280.2 | 154 | 94 | ||