Assay Details
| Gene Symbol: LOC709132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138593.2 | XP_014994079.2 | 75 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015138593.2 transcripts.
Set Membership: |
|

LOC709132
Genomic Map