Assay Details
| Gene Symbol: LOC719353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123201.2 | XP_014978687.2 | 1520 | 85 | ||
| XM_015123202.2 | XP_014978688.2 | 1207 | 85 | |||
| XM_028840266.1 | XP_028696099.1 | 861 | 85 | |||
More Information
Set Membership: |
|

LOC719353
Genomic Map