Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849299.1 | XP_028705132.1 | 4553 | 53 | ||
| XM_028849300.1 | XP_028705133.1 | 4550 | 53 | |||
More Information
Set Membership: |
|

PLEKHG4B
Genomic Map