Assay Details
| Gene Symbol: LOC100430073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147497.2 | XP_015002983.1 | 106 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015147497.2 transcripts.
Set Membership: |
|

LOC100430073
Genomic Map