Assay Details
| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109303.4 | XP_001109303.1 | 211 | 66 | ||
| XM_015134973.2 | XP_014990459.1 | 217 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM35B| Gene Symbol: TMEM35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109303.4 | XP_001109303.1 | 211 | 66 | ||
| XM_015134973.2 | XP_014990459.1 | 217 | 66 | |||
Set Membership: |
|