Assay Details
| Gene Symbol: LCE5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801782.2 | XP_002801828.2 | 196 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002801782.2 transcripts.
Set Membership: |
|

LCE5A
Genomic Map