Assay Details
| Gene Symbol: LOC716404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836627.1 | XP_028692460.1 | 7070 | 99 | ||
| XM_028836628.1 | XP_028692461.1 | 6587 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC716404| Gene Symbol: LOC716404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836627.1 | XP_028692460.1 | 7070 | 99 | ||
| XM_028836628.1 | XP_028692461.1 | 6587 | 99 | |||