Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088539.4 | XP_001088539.1 | 81 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001088539.4 transcripts.
Set Membership: |
|

AVPR2
Genomic Map