Assay Details
| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133574.2 | XP_014989060.2 | 399 | 69 | ||
| XM_015133575.2 | XP_014989061.2 | 399 | 69 | |||
| XM_015133576.2 | XP_014989062.1 | 444 | 69 | |||
More Information
Set Membership: |
|

SUN3
Genomic Map