Assay Details
| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133247.2 | XP_014988733.2 | 4259 | 95 | ||
| XM_015133248.2 | XP_014988734.2 | 826 | 95 | |||
| XM_015133249.2 | XP_014988735.2 | 780 | 95 | |||
| XM_028845112.1 | XP_028700945.1 | 617 | 95 | |||
More Information
Set Membership: |
|

ZNF316
Genomic Map