Assay Details
| Gene Symbol: LOC718458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839096.1 | XP_028694929.1 | 470 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC718458| Gene Symbol: LOC718458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839096.1 | XP_028694929.1 | 470 | 85 | ||
Set Membership: |
|