Assay Details
| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145273.2 | XP_015000759.1 | 595 | 64 | ||
| XM_028852637.1 | XP_028708470.1 | 753 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNB3| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145273.2 | XP_015000759.1 | 595 | 64 | ||
| XM_028852637.1 | XP_028708470.1 | 753 | 64 | |||
Set Membership: |
|