Assay Details
| Gene Symbol: RTL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149716.2 | XP_015005202.2 | 190 | 90 | ||
| XM_028827982.1 | XP_028683815.1 | 305 | 90 | |||
| XM_028827983.1 | XP_028683816.1 | 313 | 90 | |||
| GenBank mRNA | JV593742.1 | - | 142 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015149716.2, XM_028827982.1, XM_028827983.1 transcripts.
Set Membership: |
|

RTL10
Genomic Map