Assay Details
| Gene Symbol: PPP1R2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088324.4 | XP_001088324.1 | 823 | 110 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R2C| Gene Symbol: PPP1R2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088324.4 | XP_001088324.1 | 823 | 110 | ||
Set Membership: |
|