Assay Details
| Gene Symbol: SLC66A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131767.2 | XP_014987253.2 | 291 | 124 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC66A1L| Gene Symbol: SLC66A1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131767.2 | XP_014987253.2 | 291 | 124 | ||
Set Membership: |
|