Assay Details
| Gene Symbol: LOC107000709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150652.2 | XP_015006138.2 | 574 | 78 | ||
| XM_028827511.1 | XP_028683344.1 | 574 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107000709| Gene Symbol: LOC107000709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150652.2 | XP_015006138.2 | 574 | 78 | ||
| XM_028827511.1 | XP_028683344.1 | 574 | 78 | |||