Assay Details
| Gene Symbol: LOC107000709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150652.2 | XP_015006138.2 | 691 | 62 | ||
| XM_028827511.1 | XP_028683344.1 | 691 | 62 | |||
More Information
Set Membership: |
|

LOC107000709
Genomic Map