Assay Details
| Gene Symbol: LOC107000709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150652.2 | XP_015006138.2 | 799 | 66 | ||
| XM_028827510.1 | XP_028683343.1 | 691 | 66 | |||
More Information
Set Membership: |
|

LOC107000709
Genomic Map