Assay Details
| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144588.2 | XP_015000074.2 | 445 | 65 | ||
| XM_015144590.2 | XP_015000076.2 | 612 | 65 | |||
More Information
Set Membership: |
|

C7H14orf180
Genomic Map