Assay Details
| Gene Symbol: LOC702712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151760.2 | XP_015007246.1 | 1552 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC702712| Gene Symbol: LOC702712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151760.2 | XP_015007246.1 | 1552 | 107 | ||
Set Membership: |
|