Assay Details
| Gene Symbol: LOC713144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131121.2 | XP_014986607.2 | 1081 | 80 | ||
| XM_028844049.1 | XP_028699882.1 | 1092 | 80 | |||
More Information
Set Membership: |
|

LOC713144
Genomic Map