Assay Details
| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097428.3 | XP_001097428.1 | 1442 | 84 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPEP2NB| Gene Symbol: DPEP2NB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097428.3 | XP_001097428.1 | 1442 | 84 | ||
Set Membership: |
|