Assay Details
| Gene Symbol: LOC100429936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846550.1 | XP_028702383.1 | 163 | 89 | ||
| Gene Symbol: LOC100428559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028845611.1 | XP_028701444.1 | 95 | 89 | ||
More Information
Set Membership: |
|

LOC100429936
Genomic Map