Assay Details
| Gene Symbol: AP5S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194748.1 | NP_001181677.1 | 241 | 100 | ||
| GenBank mRNA | JU335318.1 | - | 206 | 100 | ||
| JU476272.1 | - | 200 | 100 | |||
| JV047557.1 | - | 214 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194748.1 transcripts.
Set Membership: |
|

AP5S1
Genomic Map