Assay Details
| Gene Symbol: TICAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195501.2 | NP_001182430.1 | 376 | 147 | ||
| GenBank mRNA | JU322803.1 | - | 76 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195501.2 transcripts.
Set Membership: |
|

TICAM2
Genomic Map