Assay Details
| Gene Symbol: ERRFI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261681.1 | NP_001248610.1 | 269 | 79 | ||
| GenBank mRNA | JU335057.1 | - | 316 | 79 | ||
| JV047417.1 | - | 269 | 79 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERRFI1| Gene Symbol: ERRFI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261681.1 | NP_001248610.1 | 269 | 79 | ||
| GenBank mRNA | JU335057.1 | - | 316 | 79 | ||
| JV047417.1 | - | 269 | 79 | |||