Assay Details
| Gene Symbol: TCEAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260827.1 | NP_001247756.1 | 202 | 100 | ||
| GenBank mRNA | JU334820.1 | - | 196 | 100 | ||
| JU475847.1 | - | 183 | 100 | |||
| JV047263.1 | - | 202 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260827.1 transcripts.
Set Membership: |
|

TCEAL9
Genomic Map