Assay Details
| Gene Symbol: ZBTB18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257892.1 | NP_001244821.1 | 158 | 91 | ||
| GenBank mRNA | JU320017.1 | - | 158 | 91 | ||
| JU473001.1 | - | 158 | 91 | |||
| JV492077.1 | - | 168 | 91 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257892.1 transcripts.
Set Membership: |
|

ZBTB18
Genomic Map