Assay Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261719.1 | NP_001248648.1 | 114 | 87 | ||
| GenBank mRNA | JU321502.1 | - | 353 | 87 | ||
| JU470715.1 | - | 114 | 87 | |||
| JV608972.1 | - | 363 | 87 | |||
More Information
Set Membership: |
|

SMIM13
Genomic Map