Assay Details
| Gene Symbol: RANBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194574.2 | NP_001181503.2 | 918 | 86 | ||
| GenBank mRNA | JU333979.1 | - | 887 | 86 | ||
| JU475162.1 | - | 884 | 86 | |||
More Information
Set Membership: |
|

RANBP6
Genomic Map