Assay Details
| Gene Symbol: ODC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198686.2 | NP_001185615.2 | 684 | 72 | ||
| XM_015111639.2 | XP_014967125.2 | 595 | 72 | |||
| XM_015111640.2 | XP_014967126.2 | 749 | 72 | |||
| GenBank mRNA | JU322493.1 | - | 624 | 72 | ||
| JU471203.1 | - | 618 | 72 | |||
| JV043396.1 | - | 687 | 72 | |||
| JV636834.1 | - | 776 | 72 | |||
More Information
Set Membership: |
|

ODC1
Genomic Map