Assay Details
| Gene Symbol: UQCRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198732.1 | NP_001185661.1 | 192 | 104 | ||
| XM_015140742.2 | XP_014996228.1 | 308 | 104 | |||
| GenBank mRNA | CN804540.1 | - | 192 | 104 | ||
| JU472822.1 | - | 178 | 104 | |||
| JV636100.1 | - | 215 | 104 | |||
More Information
Set Membership: |
|

UQCRQ
Genomic Map