Assay Details
| Gene Symbol: UQCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257804.1 | NP_001244733.1 | 120 | 101 | ||
| XM_028838439.1 | XP_028694272.1 | 149 | 101 | |||
| GenBank mRNA | JU332169.1 | - | 120 | 101 | ||
| JU473260.1 | - | 122 | 101 | |||
| JV045347.1 | - | 100 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257804.1, XM_028838439.1 transcripts.
Set Membership: |
|

UQCR
Genomic Map