Assay Details
| Gene Symbol: UQCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257804.1 | NP_001244733.1 | 275 | 110 | ||
| GenBank mRNA | JU332169.1 | - | 275 | 110 | ||
| JU473260.1 | - | 277 | 110 | |||
| JV045347.1 | - | 255 | 110 | |||
| JV705724.1 | - | 97 | 110 | |||
More Information
Set Membership: |
|

UQCR
Genomic Map