Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195222.1 | NP_001182151.1 | 358 | 95 | ||
| XM_015141275.2 | XP_014996761.1 | 462 | 95 | |||
| GenBank mRNA | CD766917.1 | - | 362 | 95 | ||
| JU321130.1 | - | 319 | 95 | |||
More Information
Set Membership: |
|

PTTG1
Genomic Map