Assay Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008511.2 | NP_001008511.2 | 2 - 3 | 441 | 61 | |
| XM_006230755.1 | XP_006230817.1 | 2 - 3 | 554 | 61 | ||
| GenBank mRNA | AY280663.1 | - | 1 - 2 | 335 | 61 | |
| BC104701.1 | - | 2 - 3 | 441 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esrra
Genomic Map