Assay Details
| Gene Symbol: Esrra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008511.2 | NP_001008511.2 | 5 - 6 | 855 | 77 | |
| XM_006230755.1 | XP_006230817.1 | 5 - 6 | 968 | 77 | ||
| GenBank mRNA | AY280663.1 | - | 4 - 5 | 749 | 77 | |
| BC104701.1 | - | 5 - 6 | 855 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esrra
Genomic Map