Assay Details
| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017144.2 | NP_058840.1 | 4 - 5 | 257 | 61 | |
| GenBank mRNA | DQ062462.1 | - | 2 - 3 | 153 | 61 | |
| M57679.1 | - | 4 - 5 | 240 | 61 | ||
| M92074.1 | - | 3 - 4 | 207 | 61 | ||
| X58499.1 | - | 4 - 5 | 258 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnni3
Genomic Map