Assay Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031764.3 | NP_113952.2 | 1 - 2 | 87 | 64 | |
| XM_006250226.2 | XP_006250288.1 | 2 - 3 | 3384 | 64 | ||
| XM_006250227.2 | XP_006250289.1 | 4 - 5 | 719 | 64 | ||
| XM_006250228.2 | XP_006250290.1 | 3 - 4 | 500 | 64 | ||
| XM_006250229.2 | XP_006250291.1 | 3 - 4 | 448 | 64 | ||
| XM_008769761.1 | XP_008767983.1 | 3 - 4 | 462 | 64 | ||
| XM_008769762.1 | XP_008767984.1 | 3 - 4 | 394 | 64 | ||
| XM_008769763.1 | XP_008767985.1 | 3 - 4 | 700 | 64 | ||
| GenBank mRNA | BC161961.1 | - | 1 - 2 | 87 | 9027893 | 64 |
| EV772283.1 | - | 1 - 2 | 127 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddr2
Genomic Map