Assay Details
Gene Symbol: Selm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115013.1 | NP_001108485.1 | 3 - 4 | 247 | 69 | |
GenBank mRNA | BI282694.1 | - | 2 - 3 | 69 | ||
CB607044.1 | - | 3 - 4 | 219 | 69 | ||
FQ223498.1 | - | 3 - 4 | 202 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
