Assay Details
| Gene Symbol: LOC303566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247329.2 | XP_006247391.1 | 2 - 3 | 295 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC303566| Gene Symbol: LOC303566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247329.2 | XP_006247391.1 | 2 - 3 | 295 | 86 | |
Set Membership: |
|