Assay Details
| Gene Symbol: LOC100910990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235456.2 | XP_006235518.1 | 1 - 2 | 167 | 57 | |
| XM_008762412.1 | XP_008760634.1 | 1 - 2 | 171 | 57 | ||
| Gene Symbol: Rbm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762382.1 | XP_008760604.1 | 1 - 2 | 171 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235456.2, XM_008762382.1, XM_008762412.1 transcripts.
Set Membership: |
|

LOC100910990
Genomic Map