Assay Details
| Gene Symbol: Dtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108594.1 | NP_001102064.1 | 3 - 4 | 439 | 80 | |
| XM_006235128.2 | XP_006235190.1 | 3 - 4 | 479 | 80 | ||
| GenBank mRNA | BC159413.1 | - | 3 - 4 | 422 | 8367451 | 80 |
| FQ214547.1 | - | 3 - 4 | 411 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dtd1
Genomic Map