Assay Details
| Gene Symbol: Smoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002835.1 | NP_001002835.1 | 10 - 11 | 1044 | 129 | |
| XM_006240284.2 | XP_006240346.1 | 10 - 11 | 1350 | 129 | ||
| XM_006240285.2 | XP_006240347.1 | 10 - 11 | 1350 | 129 | ||
| XM_008764785.1 | XP_008763007.1 | 11 - 12 | 1450 | 129 | ||
| XM_008764786.1 | XP_008763008.1 | 11 - 12 | 1417 | 129 | ||
| GenBank mRNA | FQ222878.1 | - | 10 - 11 | 1301 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smoc1
Genomic Map